Anti-CD62L Rat Monoclonal Antibody [clone: MEL-14]

Supplier: Abcam
AB119834-1MG AB119834-100UG AB119834-20UG
ABCA_AB119834-1MGEA 11108.98 CAD
ABCA_AB119834-1MG ABCA_AB119834-100U ABCA_AB119834-20UG
Anti-CD62L Rat Monoclonal Antibody [clone: MEL-14]
Antibodies
Rat monoclonal [MEL-14] to CD62L.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: CDK1
Clonality: Monoclonal
Clone: YE324
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Rat; Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR