Anti-Histone H2B (acetyl K12 + K12 + K16 + K23 + K23 + K23 + K23 + K23 + K5 + K7) Mouse Monoclonal Antibody [clone: mAbcam 52484]

Supplier: Abcam
AB237964-100UG AB237964-1MG
ABCA_AB237964-100UEA 1791.72 CAD
ABCA_AB237964-100U ABCA_AB237964-1MG
Anti-Histone H2B (acetyl K12 + K12 + K16 + K23 + K23 + K23 + K23 + K23 + K5 + K7) Mouse Monoclonal Antibody [clone: mAbcam 52484]
Antibodies
Mouse monoclonal [mAbcam 52484] to Histone H2B - ChIP Grade – BSA and Azide free.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Histone H2B
Clonality: Monoclonal
Clone: mAbcam 52484
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR