Anti-SIRP alpha Mouse Monoclonal Antibody [clone: OX-41]

Supplier: Abcam
AB307563-20UG AB307563-1MG AB307563-100UG AB307563-10UG
ABCA_AB307563-20UGEA 436.08 CAD
ABCA_AB307563-20UG ABCA_AB307563-1MG ABCA_AB307563-100U ABCA_AB307563-10UG
Anti-SIRP alpha Mouse Monoclonal Antibody [clone: OX-41]
Antibodies
Mouse monoclonal [OX-41] to SIRP alpha.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: SIRP alpha
Clonality: Monoclonal
Clone: OX-41
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR