Anti-GABA Transporter 1 / GAT 1 Rabbit Monoclonal Antibody [clone: EPR12998]

Supplier: Abcam
AB177483-100UL AB177483-1ML
ABCA_AB177483-100UEA 1079.14 CAD
ABCA_AB177483-100U ABCA_AB177483-1ML
Anti-GABA Transporter 1 / GAT 1 Rabbit Monoclonal Antibody [clone: EPR12998]
Antibodies
Rabbit monoclonal [EPR12998] to GABA Transporter 1 / GAT 1.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: GABA Transporter 1 / GAT 1
Clonality: Monoclonal
Clone: EPR12998
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR