Anti-Histone H4 (acetyl K5) Rabbit Monoclonal Antibody [clone: EP1000Y]

Supplier: Abcam
AB51997-100UL AB51997-1ML
ABCA_AB51997-100ULEA 1406.2 CAD
ABCA_AB51997-100UL ABCA_AB51997-1ML
Anti-Histone H4 (acetyl K5) Rabbit Monoclonal Antibody [clone: EP1000Y]
Antibodies
Rabbit monoclonal [EP1000Y] to Histone H4 (acetyl K5) - ChIP Grade.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Histone H4
Clonality: Monoclonal
Clone: EP1000Y
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse;Rat;Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR