Anti-PI 3 Kinase catalytic subunit gamma/PI3K-gamma Rabbit Monoclonal Antibody [clone: EPR25156-60]

Supplier: Abcam
AB302958-1ML AB302958-100UL AB302958-20UL
ABCA_AB302958-1MLEA 11564.02 CAD
ABCA_AB302958-1ML ABCA_AB302958-100U ABCA_AB302958-20UL
Anti-PI 3 Kinase catalytic subunit gamma/PI3K-gamma Rabbit Monoclonal Antibody [clone: EPR25156-60]
Antibodies
Rabbit monoclonal [EPR25156-60] to PI 3 Kinase catalytic subunit gamma/PI3K-gamma.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: PI 3 Kinase catalytic subunit gamma/PI3K-gamma
Clonality: Monoclonal
Clone: EPR25156-60
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR