Anti-Polyethylene glycol Rabbit Monoclonal Antibody [clone: EPR21993-154]

Supplier: Abcam
AB256481-1MG AB256481-100UG
ABCA_AB256481-1MGEA 16158.66 CAD
ABCA_AB256481-1MG ABCA_AB256481-100U
Anti-Polyethylene glycol Rabbit Monoclonal Antibody [clone: EPR21993-154]
Antibodies
Rabbit monoclonal [EPR21993-154] to Polyethylene glycol - BSA and Azide free.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Polyethylene glycol
Clonality: Monoclonal
Clone: EPR21993-154
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR