Anti-FGF 23 Rabbit Monoclonal Antibody [clone: EPR25309-57]

Supplier: Abcam
AB307420-10UL AB307420-20UL AB307420-100UL AB307420-1ML
ABCA_AB307420-10ULEA 425.02 CAD
ABCA_AB307420-10UL ABCA_AB307420-20UL ABCA_AB307420-100U ABCA_AB307420-1ML
Anti-FGF 23 Rabbit Monoclonal Antibody [clone: EPR25309-57]
Antibodies
Rabbit monoclonal [EPR25309-57] to FGF 23.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: FGF 23
Clonality: Monoclonal
Clone: EPR25309-57
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR