Anti-Tyrosine Hydroxylase Rabbit Recombinant Multiclonal Antibody [clone: RM1085]

Supplier: Abcam
AB315253-100UG AB315253-1MG
ABCA_AB315253-100UEA 1791.72 CAD
ABCA_AB315253-100U ABCA_AB315253-1MG
Anti-Tyrosine Hydroxylase Rabbit Recombinant Multiclonal Antibody [clone: RM1085]
Antibodies
Rabbit recombinant multiclonal [RM1085] to Tyrosine Hydroxylase - BSA and Azide free.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Tyrosine Hydroxylase
Clonality: Recombinant Multiclonal
Clone: RM1085
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR