Anti-PYGL (phospho S430) + PYGM (phospho S430) (phospho S430) Rabbit Monoclonal Antibody [clone: EPR20852-26]

Supplier: Abcam
AB314428-20UL AB314428-1ML AB314428-100UL
ABCA_AB314428-20ULEA 436.08 CAD
ABCA_AB314428-20UL ABCA_AB314428-1ML ABCA_AB314428-100U
Anti-PYGL (phospho S430) + PYGM (phospho S430) (phospho S430) Rabbit Monoclonal Antibody [clone: EPR20852-26]
Antibodies
Rabbit monoclonal [EPR20852-26] to PYGL (phospho S430) + PYGM (phospho S430).

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: PYGL (phospho S430) + PYGM (phospho S430)
Clonality: Monoclonal
Clone: EPR20852-26
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR