Anti-Sodium Potassium ATPase Rat Monoclonal Antibody [clone: EP1845Y]

Supplier: Abcam
AB283341-100UG AB283341-1MG
ABCA_AB283341-100UEA 1791.72 CAD
ABCA_AB283341-100U ABCA_AB283341-1MG
Anti-Sodium Potassium ATPase Rat Monoclonal Antibody [clone: EP1845Y]
Antibodies
Rat monoclonal [EP1845Y] to Sodium Potassium ATPase - Chimeric – BSA and Azide free.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Sodium Potassium ATPase
Clonality: Monoclonal
Clone: EP1845Y
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR