Anti-Keratin 12/K12 Rabbit Monoclonal Antibody [clone: EPR1609(2)]

Supplier: Abcam
AB124975-1ML AB124975-100UL
ABCA_AB124975-1MLEA 7892.1 CAD
ABCA_AB124975-1ML ABCA_AB124975-100U
Anti-Keratin 12/K12 Rabbit Monoclonal Antibody [clone: EPR1609(2)]
Antibodies
Rabbit monoclonal [EPR1609(2)] to Keratin 12/K12.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Keratin 12/K12
Clonality: Monoclonal
Clone: EPR1609(2)
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR