Anti-ICOS Ligand/ICOSL Rabbit Monoclonal Antibody [clone: EPR20240-231]

Supplier: Abcam
AB272761-1MG AB272761-100UG
ABCA_AB272761-1MGEA 15534.56 CAD
ABCA_AB272761-1MG ABCA_AB272761-100U
Anti-ICOS Ligand/ICOSL Rabbit Monoclonal Antibody [clone: EPR20240-231]
Antibodies
Rabbit monoclonal [EPR20240-231] to ICOS Ligand/ICOSL - BSA and Azide free (Capture).

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: ICOS Ligand/ICOSL
Clonality: Monoclonal
Clone: EPR20240-231
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR