Anti-PCNA Mouse Monoclonal Antibody [clone: 24/PCNA]

Supplier: Abcam
AB280088-20UG AB280088-100UG AB280088-1MG
ABCA_AB280088-20UGEA 276.5 CAD
ABCA_AB280088-20UG ABCA_AB280088-100U ABCA_AB280088-1MG
Anti-PCNA Mouse Monoclonal Antibody [clone: 24/PCNA]
Antibodies
Mouse monoclonal [24/PCNA] to PCNA.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: PCNA
Clonality: Monoclonal
Clone: 24/PCNA
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR