Anti-Fibronectin Mouse Monoclonal Antibody [clone: 10/Fibronectin]

Supplier: Abcam
AB281575-20UG AB281575-1MG AB281575-100UG
ABCA_AB281575-20UGEA 276.5 CAD
ABCA_AB281575-20UG ABCA_AB281575-1MG ABCA_AB281575-100U
Anti-Fibronectin Mouse Monoclonal Antibody [clone: 10/Fibronectin]
Antibodies
Mouse monoclonal [10/Fibronectin] to Fibronectin.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Fibronectin
Clonality: Monoclonal
Clone: 10/Fibronectin
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR