Anti-Dopamine Transporter Rabbit Monoclonal Antibody [clone: EPR19695] (Alexa Fluor® 568)

Supplier: Abcam
AB303466-10UL AB303466-100UL
ABCA_AB303466-10ULEA 336.54 CAD
ABCA_AB303466-10UL ABCA_AB303466-100U
Anti-Dopamine Transporter Rabbit Monoclonal Antibody [clone: EPR19695] (Alexa Fluor® 568)
Antibodies
Alexa Fluor® 568 Rabbit monoclonal [EPR19695] to Dopamine Transporter.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Dopamine Transporter
Clonality: Monoclonal
Clone: EPR19695
Conjugation: Alexa Fluor® 568
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR