Anti-PKC delta Mouse Monoclonal Antibody [clone: 14/PKC delta]

Supplier: Abcam
AB307233-100UG AB307233-20UG AB307233-1MG AB307233-10UG
ABCA_AB307233-100UEA 1257.68 CAD
ABCA_AB307233-100U ABCA_AB307233-20UG ABCA_AB307233-1MG ABCA_AB307233-10UG
Anti-PKC delta Mouse Monoclonal Antibody [clone: 14/PKC delta]
Antibodies
Mouse monoclonal [14/PKC delta] to PKC delta.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type:
Antigen: ALB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Pigeon
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR