Anti-GABA A Receptor alpha 5/GABRA5 Mouse Monoclonal Antibody [clone: N415--24]

Supplier: Abcam
AB302962-100UG AB302962-1MG AB302962-10UG AB302962-20UG
ABCA_AB302962-100UEA 1257.68 CAD
ABCA_AB302962-100U ABCA_AB302962-1MG ABCA_AB302962-10UG ABCA_AB302962-20UG
Anti-GABA A Receptor alpha 5/GABRA5 Mouse Monoclonal Antibody [clone: N415--24]
Antibodies
Mouse monoclonal [N415/24] to GABA A Receptor alpha 5/GABRA5.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: GABA A Receptor alpha 5/GABRA5
Clonality: Monoclonal
Clone: N415--24
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR