Anti-Histone H2B Rabbit Monoclonal Antibody [clone: EPR27299-85]

Supplier: Abcam
AB309100-1ML AB309100-100UL AB309100-20UL
ABCA_AB309100-1MLEA 11287.52 CAD
ABCA_AB309100-1ML ABCA_AB309100-100U ABCA_AB309100-20UL
Anti-Histone H2B Rabbit Monoclonal Antibody [clone: EPR27299-85]
Antibodies
Rabbit monoclonal [EPR27299-85] to Histone H2B (mutated E76Q).

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Histone H2B
Clonality: Monoclonal
Clone: EPR27299-85
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR