Anti-Fyn Mouse Monoclonal Antibody [clone: 25/FYN H1L1]

Supplier: Abcam
AB300634-20UG AB300634-100UG AB300634-1MG
ABCA_AB300634-20UGEA 436.08 CAD
ABCA_AB300634-20UG ABCA_AB300634-100U ABCA_AB300634-1MG
Anti-Fyn Mouse Monoclonal Antibody [clone: 25/FYN H1L1]
Antibodies
Mouse monoclonal [25/FYN] to Fyn.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Fyn
Clonality: Monoclonal
Clone: 25/FYN H1L1
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR