Anti-SIGLEC8 Rabbit Monoclonal Antibody [clone: EPR24419-56]

Supplier: Abcam
AB305297-100UL AB305297-20UL AB305297-1ML
ABCA_AB305297-100UEA 1257.68 CAD
ABCA_AB305297-100U ABCA_AB305297-20UL ABCA_AB305297-1ML
Anti-SIGLEC8 Rabbit Monoclonal Antibody [clone: EPR24419-56]
Antibodies
Rabbit monoclonal [EPR24419-56] to SIGLEC8.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences,More Product Information
New Product
Order Now

SPECIFICATIONS

Antigen symbol SIGLEC8
Antigen name SAF2
Environmentally Preferable
Antigen synonyms Sialic acid-binding Ig-like lectin 8|Siglec-8|Sialoadhesin family member 2|SAF-2
Antibody type Primary
Clonality Monoclonal
Clone EPR24419-56
Reactivity
Host Rabbit
Isotype IgG
Western blot Yes
ImmunoChemistry Yes
ImmunoPrecipitation Yes
ImmunoFluorescence Yes
Flow cytometry Yes
Immunogen The exact immunogen used to generate this antibody is proprietary information
Form Liquid
Molecular weight 54042 Da
Purification Protein A affinity purification
Storage temperature Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C. Avoid freeze/thaw cycle.
Shipping temperature 4 °C
Tested applications Flow Cyt (Intra), ICC/IF

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR