Anti-Cytokeratin 19 Mouse Monoclonal Antibody [clone: LP2K]

Supplier: Abcam
AB255939-100UG AB255939-1MG
ABCA_AB255939-100UEA 1791.72 CAD
ABCA_AB255939-100U ABCA_AB255939-1MG
Anti-Cytokeratin 19 Mouse Monoclonal Antibody [clone: LP2K]
Antibodies
Mouse monoclonal [LP2K] to Cytokeratin 19 - BSA and Azide free.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Cytokeratin 19
Clonality: Monoclonal
Clone: LP2K
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR