Anti-pro Caspase-3 Rabbit Monoclonal Antibody [clone: E83-103]

Supplier: Abcam
AB32499-100UL AB32499-20UL AB32499-1ML
ABCA_AB32499-100ULEA 1316.14 CAD
ABCA_AB32499-100UL ABCA_AB32499-20UL ABCA_AB32499-1ML
Anti-pro Caspase-3 Rabbit Monoclonal Antibody [clone: E83-103]
Antibodies
Rabbit monoclonal [E83-103] to pro Caspase-3.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: pro Caspase-3
Clonality: Monoclonal
Clone: E83-103
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse;Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR