Anti-Claudin 1 Rabbit Recombinant Multiclonal Antibody [clone: RM1186]

Supplier: Abcam
AB317709-20UL AB317709-100UL AB317709-1ML
ABCA_AB317709-20ULEA 366.56 CAD
ABCA_AB317709-20UL ABCA_AB317709-100U ABCA_AB317709-1ML
Anti-Claudin 1 Rabbit Recombinant Multiclonal Antibody [clone: RM1186]
Antibodies
Rabbit recombinant multiclonal [RM1186] to Claudin 1.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Claudin 1
Clonality: Recombinant Multiclonal
Clone: RM1186
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR