Anti-Serotonin transporter Rabbit Monoclonal Antibody [clone: EPR23530-3]

Supplier: Abcam
AB254358-1ML AB254358-20UL AB254358-100UL
ABCA_AB254358-1MLEA 10832.48 CAD
ABCA_AB254358-1ML ABCA_AB254358-20UL ABCA_AB254358-100U
Anti-Serotonin transporter Rabbit Monoclonal Antibody [clone: EPR23530-3]
Antibodies
Rabbit monoclonal [EPR23530-3] to Serotonin transporter.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Serotonin transporter
Clonality: Monoclonal
Clone: EPR23530-3
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR