Anti-Oncostatin M/OSM Rabbit Monoclonal Antibody [clone: EPR26065-14]

Supplier: Abcam
AB307951-1ML AB307951-20UL AB307951-100UL
ABCA_AB307951-1MLEA 11287.52 CAD
ABCA_AB307951-1ML ABCA_AB307951-20UL ABCA_AB307951-100U
Anti-Oncostatin M/OSM Rabbit Monoclonal Antibody [clone: EPR26065-14]
Antibodies
Rabbit monoclonal [EPR26065-14] to Oncostatin M/OSM.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Oncostatin M/OSM
Clonality: Monoclonal
Clone: EPR26065-14
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR