Anti-IgE Rabbit Monoclonal Antibody [clone: EPR26777-14]

Supplier: Abcam
AB309141-100UL AB309141-1ML AB309141-20UL
ABCA_AB309141-100UEA 1257.68 CAD
ABCA_AB309141-100U ABCA_AB309141-1ML ABCA_AB309141-20UL
Anti-IgE Rabbit Monoclonal Antibody [clone: EPR26777-14]
Antibodies
Rabbit monoclonal [EPR26777-14] to IgE.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: PITX2/RGS
Clonality: Monoclonal
Clone: EPR22417
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR