Anti-Metabotropic Glutamate Receptor 7/MGLUR7 Rabbit Monoclonal Antibody [clone: EPR24530-89]

Supplier: Abcam
AB302784-1ML AB302784-100UL AB302784-20UL
ABCA_AB302784-1MLEA 11564.02 CAD
ABCA_AB302784-1ML ABCA_AB302784-100U ABCA_AB302784-20UL
Anti-Metabotropic Glutamate Receptor 7/MGLUR7 Rabbit Monoclonal Antibody [clone: EPR24530-89]
Antibodies
Rabbit monoclonal [EPR24530-89] to Metabotropic Glutamate Receptor 7/MGLUR7.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Metabotropic Glutamate Receptor 7/MGLUR7
Clonality: Monoclonal
Clone: EPR24530-89
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR