Anti-hnRNP A1 (citrulline R140) Rabbit Monoclonal Antibody [clone: EPR20178]

Supplier: Abcam
AB251488-100UG AB251488-1MG
ABCA_AB251488-100UEA 1791.72 CAD
ABCA_AB251488-100U ABCA_AB251488-1MG
Anti-hnRNP A1 (citrulline R140) Rabbit Monoclonal Antibody [clone: EPR20178]
Antibodies
Rabbit monoclonal [EPR20178] to hnRNP A1 (citrulline R140) - BSA and Azide free.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: hnRNP A1
Clonality: Monoclonal
Clone: EPR20178
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Recombinant full length protein
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR