Anti-GLUT1 Rabbit Polyclonal Antibody

Supplier: Abcam
AB15309-100UL AB15309-500UL
ABCA_AB15309-100ULEA 1613.18 CAD
ABCA_AB15309-100UL ABCA_AB15309-500UL
Anti-GLUT1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to Glucose Transporter GLUT1.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: GLUT1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR