Anti-2 Hydroxy phytanoyl CoA lyase/HACL1 Rabbit Monoclonal Antibody [clone: EPR16812]

Supplier: Abcam
AB197025-1ML AB197025-100UL
ABCA_AB197025-1MLEA 9089.74 CAD
ABCA_AB197025-1ML ABCA_AB197025-100U
Anti-2 Hydroxy phytanoyl CoA lyase/HACL1 Rabbit Monoclonal Antibody [clone: EPR16812]
Antibodies
Rabbit monoclonal [EPR16812] to 2 Hydroxy phytanoyl CoA lyase/HACL1.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: 2 Hydroxy phytanoyl CoA lyase/HACL1
Clonality: Monoclonal
Clone: EPR16812
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR