Anti-Hsp60 Mouse Monoclonal Antibody [clone: 24/HSP60 H1L1]

Supplier: Abcam
AB300659-20UG AB300659-10UG AB300659-1MG AB300659-100UG
ABCA_AB300659-20UGEA 276.5 CAD
ABCA_AB300659-20UG ABCA_AB300659-10UG ABCA_AB300659-1MG ABCA_AB300659-100U
Anti-Hsp60 Mouse Monoclonal Antibody [clone: 24/HSP60 H1L1]
Antibodies
Mouse monoclonal [24/HSP60] to Hsp60.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Hsp60
Clonality: Monoclonal
Clone: 24/HSP60 H1L1
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human; Mouse; Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR