Anti-Serine/threonine-protein kinase 4/MST-1 Rabbit Monoclonal Antibody [clone: EP1465Y]

Supplier: Abcam
AB51134-100UL AB51134-1ML
ABCA_AB51134-100ULEA 1069.66 CAD
ABCA_AB51134-100UL ABCA_AB51134-1ML
Anti-Serine/threonine-protein kinase 4/MST-1 Rabbit Monoclonal Antibody [clone: EP1465Y]
Antibodies
Rabbit monoclonal [EP1465Y] to Serine/threonine-protein kinase 4/MST-1.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Serine/threonine-protein kinase 4/MST-1
Clonality: Monoclonal
Clone: EP1465Y
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR