Anti-IgG Goat Polyclonal Antibody (Gold, 20nm)

Supplier: Abcam
AB270563-200UL AB270563-1ML
ABCA_AB270563-200UEA 336.54 CAD
ABCA_AB270563-200U ABCA_AB270563-1ML
Anti-IgG Goat Polyclonal Antibody (Gold, 20nm)
Antibodies
Goat Anti-Rabbit IgG (20nm Gold, 10 OD).

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Gold, 20nm
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR