Anti-PORCN/PPN Rabbit Monoclonal Antibody [clone: EPR19741]

Supplier: Abcam
AB201793-1ML AB201793-100UL
ABCA_AB201793-1MLEA 8811.66 CAD
ABCA_AB201793-1ML ABCA_AB201793-100U
Anti-PORCN/PPN Rabbit Monoclonal Antibody [clone: EPR19741]
Antibodies
Rabbit monoclonal [EPR19741] to PORCN/PPN.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: PORCN/PPN
Clonality: Monoclonal
Clone: EPR19741
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR