Anti-Glucose 6 Phosphate Dehydrogenase Rabbit Monoclonal Antibody [clone: EPR20668]

Supplier: Abcam
AB231828-1MG AB231828-100UG
ABCA_AB231828-1MGEA 16158.66 CAD
ABCA_AB231828-1MG ABCA_AB231828-100U
Anti-Glucose 6 Phosphate Dehydrogenase Rabbit Monoclonal Antibody [clone: EPR20668]
Antibodies
Rabbit monoclonal [EPR20668] to Glucose 6 Phosphate Dehydrogenase - BSA and Azide free.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Glucose 6 Phosphate Dehydrogenase
Clonality: Monoclonal
Clone: EPR20668
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR