Anti-Cytokeratin 14 Rabbit Monoclonal Antibody [clone: EPR17336]

Supplier: Abcam
AB197893-1ML AB197893-100UL
ABCA_AB197893-1MLEA 8257.08 CAD
ABCA_AB197893-1ML ABCA_AB197893-100U
Anti-Cytokeratin 14 Rabbit Monoclonal Antibody [clone: EPR17336]
Antibodies
Rabbit monoclonal [EPR17336] to Cytokeratin 14.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Cytokeratin 14
Clonality: Monoclonal
Clone: EPR17336
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR