Anti-CXCL11 Rabbit Monoclonal Antibody [clone: EPR23814-16]

Supplier: Abcam
AB259863-1ML AB259863-20UL AB259863-100UL
ABCA_AB259863-1MLEA 11753.62 CAD
ABCA_AB259863-1ML ABCA_AB259863-20UL ABCA_AB259863-100U
Anti-CXCL11 Rabbit Monoclonal Antibody [clone: EPR23814-16]
Antibodies
Rabbit monoclonal [EPR23814-16] to CXCL11.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: CXCL11
Clonality: Monoclonal
Clone: EPR23814-16
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR