Anti-Carbonic Anhydrase 14/CA14 Rabbit Monoclonal Antibody [clone: EPR14307]

Supplier: Abcam
AB184180-1ML AB184180-100UL
ABCA_AB184180-1MLEA 9732.8 CAD
ABCA_AB184180-1ML ABCA_AB184180-100U
Anti-Carbonic Anhydrase 14/CA14 Rabbit Monoclonal Antibody [clone: EPR14307]
Antibodies
Rabbit monoclonal [EPR14307] to Carbonic Anhydrase 14/CA14.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Carbonic Anhydrase 14/CA14
Clonality: Monoclonal
Clone: EPR14307
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR