Anti-MBP Rat Monoclonal Antibody [clone: 12]

Supplier: Abcam
AB277483-1MG AB277483-100UG
ABCA_AB277483-1MGEA 16158.66 CAD
ABCA_AB277483-1MG ABCA_AB277483-100U
Anti-MBP Rat Monoclonal Antibody [clone: 12]
Antibodies
Rat monoclonal [12] to Myelin Basic Protein - BSA and Azide free.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: MBP
Clonality: Monoclonal
Clone: 12
Conjugation:
Epitope: Amino acids 82-87 (DENPVV).
Host: Rat
Isotype: IgG2a
Reactivity: Mouse;Rat;Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR