Anti-COX2 / Cyclooxygenase 2 Mouse Monoclonal Antibody [clone: 33/Cox-2 H1L1]

Supplier: Abcam
AB300668-1MG AB300668-10UG AB300668-20UG AB300668-100UG
ABCA_AB300668-1MGEA 7128.96 CAD
ABCA_AB300668-1MG ABCA_AB300668-10UG ABCA_AB300668-20UG ABCA_AB300668-100U
Anti-COX2 / Cyclooxygenase 2 Mouse Monoclonal Antibody [clone: 33/Cox-2 H1L1]
Antibodies
Mouse monoclonal [33/Cox-2] to COX2 / Cyclooxygenase 2.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: COX2 / Cyclooxygenase 2
Clonality: Monoclonal
Clone: 33/Cox-2 H1L1
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human; Mouse; Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR