Anti-non-muscle Myosin IIB/MYH10 Rat Monoclonal Antibody [clone: EPR22564-23]

Supplier: Abcam
AB300647-1ML AB300647-100UL AB300647-20UL AB300647-10UL
ABCA_AB300647-1MLEA 7128.96 CAD
ABCA_AB300647-1ML ABCA_AB300647-100U ABCA_AB300647-20UL ABCA_AB300647-10UL
Anti-non-muscle Myosin IIB/MYH10 Rat Monoclonal Antibody [clone: EPR22564-23]
Antibodies
Rat monoclonal [EPR22564-23] to non-muscle Myosin IIB/MYH10 - Chimeric.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: non-muscle Myosin IIB/MYH10
Clonality: Monoclonal
Clone: EPR22564-23
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR