Anti-JAK3 Rabbit Monoclonal Antibody [clone: JAK3Y980981-E10] (APC (Allophycocyanin))

Supplier: Abcam
AB278667-10TEST AB278667-100TEST
ABCA_AB278667-10TEEA 524.56 CAD
ABCA_AB278667-10TE ABCA_AB278667-100T
Anti-JAK3 Rabbit Monoclonal Antibody [clone: JAK3Y980981-E10] (APC (Allophycocyanin))
Antibodies
APC Rabbit monoclonal [JAK3Y980981-E10] to JAK3 (phospho Y980 + Y981).

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: JAK3
Clonality: Monoclonal
Clone: JAK3Y980981-E10
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR