Anti-Histone H3 (acetyl K27) Mouse Monoclonal Antibody [clone: EPR16601]

Supplier: Abcam
AB317800-100UG AB317800-1MG
ABCA_AB317800-100UEA 1791.72 CAD
ABCA_AB317800-100U ABCA_AB317800-1MG
Anti-Histone H3 (acetyl K27) Mouse Monoclonal Antibody [clone: EPR16601]
Antibodies
Mouse monoclonal [EPR16601] to Histone H3 (tri methyl K9) - Chimeric – BSA and Azide free.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Histone H3
Clonality: Monoclonal
Clone: EPR16601
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human;Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR