Anti-IgG Goat Polyclonal Antibody (Gold)

Supplier: Abcam
AB270700-200UL AB270700-1ML
ABCA_AB270700-200UEA 376.04 CAD
ABCA_AB270700-200U ABCA_AB270700-1ML
Anti-IgG Goat Polyclonal Antibody (Gold)
Antibodies
Goat Anti-Rabbit IgG (80nm Gold, 10 OD).

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Gold
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR