Anti-Influenza B Virus Nucleoprotein Rabbit Antibody [clone: EPR28205-27]

Supplier: Abcam
AB313885-1ML AB313885-100UL AB313885-20UL
ABCA_AB313885-1MLEA 11287.52 CAD
ABCA_AB313885-1ML ABCA_AB313885-100U ABCA_AB313885-20UL
Anti-Influenza B Virus Nucleoprotein Rabbit Antibody [clone: EPR28205-27]
Antibodies
Rabbit monoclonal [EPR28205-27] to Influenza B Virus Nucleoprotein.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Influenza B Virus Nucleoprotein
Clonality:
Clone: EPR28205-27
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: InfB
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR