Anti-Superoxide Dismutase 3/EC-SOD Rabbit Monoclonal Antibody [clone: EPR10860]

Supplier: Abcam
AB171738-100UL AB171738-1ML
ABCA_AB171738-100UEA 1088.62 CAD
ABCA_AB171738-100U ABCA_AB171738-1ML
Anti-Superoxide Dismutase 3/EC-SOD Rabbit Monoclonal Antibody [clone: EPR10860]
Antibodies
Rabbit monoclonal [EPR10860] to Superoxide Dismutase 3/EC-SOD.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Superoxide Dismutase 3/EC-SOD
Clonality: Monoclonal
Clone: EPR10860
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR