Anti-Glypican 3 Rabbit Monoclonal Antibody [clone: 24] FITC (Fluorescein Isothiocyanate)

Supplier: Abcam
AB275696-100TEST AB275696-25TEST
ABCA_AB275696-100TEA 1099.68 CAD
ABCA_AB275696-100T ABCA_AB275696-25TE
Anti-Glypican 3 Rabbit Monoclonal Antibody [clone: 24] FITC (Fluorescein Isothiocyanate)
Antibodies
FITC Rabbit monoclonal [024] to Glypican 3.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Glypican 3
Clonality: Monoclonal
Clone: 24
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR