Anti-WT1 Rabbit Monoclonal Antibody [clone: SP321]

Supplier: Abcam
AB224806-1ML AB224806-100UL AB224806-500UL
ABCA_AB224806-1MLEA 1712.72 CAD
ABCA_AB224806-1ML ABCA_AB224806-100U ABCA_AB224806-500U
Anti-WT1 Rabbit Monoclonal Antibody [clone: SP321]
Antibodies
Rabbit monoclonal [SP321] to Wilms Tumor Protein.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: WT1
Clonality: Monoclonal
Clone: SP321
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR