Anti-Histone H3 (acetyl K4 + K9 + K14 + K18 + K23 + K27) Rabbit Recombinant Multiclonal Antibody [clone: RM1045]

Supplier: Abcam
AB300641-1ML AB300641-100UL AB300641-20UL
ABCA_AB300641-1MLEA 9268.28 CAD
ABCA_AB300641-1ML ABCA_AB300641-100U ABCA_AB300641-20UL
Anti-Histone H3 (acetyl K4 + K9 + K14 + K18 + K23 + K27) Rabbit Recombinant Multiclonal Antibody [clone: RM1045]
Antibodies
Rabbit recombinant multiclonal [RM1045] to Histone H3 (acetyl K4 + K9 + K14 + K18 + K23 + K27).

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Histone H3
Clonality: Recombinant Multiclonal
Clone: RM1045
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR